Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG-3 Antibody (L4-PL33) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$241.50 - $564.50
Specifications
Antigen | LAG-3 |
---|---|
Clone | L4-PL33 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), NeutralizationSpecifications
LAG-3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Primate | |
17b4, 17b4 lag3, 17b4 neutralizing, anti human lag3, Anti-LAG-3, CD223, CD223 Antigen, Human LAG3, Human LAG-3, LAG3, LAG-3, LAG-3 17b4, LAG-3 blocking, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 | |
This LAG-3 Antibody (L4-PL33) was prepared from synthetic peptide corresponding to 30 aa (GPPAAAPGHPLAPGPHPAAPSSWGPRPRRY) from the first N-terminal D1 domain of human LAG-3 (lymphocyte activation gene-3). | |
Protein A or G purified |
L4-PL33 | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cancer, Immunology, Innate Immunity | |
PBS, 10% glycerol | |
3902 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title