Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG-3 Antibody (OTI8F6), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271112AF750
Description
LAG-3 Monoclonal specifically detects LAG-3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LAG-3 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
LAG3 | |
Recombinant protein fragment corresponding to amino acids 66-332 of human LAG-3 (NP_002277) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI8F6 | |
Western Blot, Immunohistochemistry | |
CD223, CD223 antigen, FDC, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
Mouse | |
Affinity Purified | |
Adaptive Immunity, Immunology, Innate Immunity | |
3902.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction