Learn More
Description
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
| Antigen | LAG-3 (CD223) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | T47-530 |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC |
| Host Species | Mouse |
| Immunogen | Human LAG-3 Recombinant Protein |
| Quantity | 25 Tests |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.