Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG3 Monoclonal Antibody (OTI8G6), TrueMAB™, OriGene
SDP

Catalog No. 501681425
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1425 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1425 Supplier OriGene Supplier No. TA807180
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD-223 (LAG-3, Lymphocyte-activation protein 3) is a 70 kDa surface glycoprotein belonging to the Ig superfamily. CD-223 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of CD-223 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express CD-223 at significantly higher levels than CD4+ T cells. Coexpression of CD-223 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells). LAG-3 contains 4 extracellular Ig-like domains and its gene contains 8 exons. The sequence data, exon/intron organization, and chromosomal localization all indicate homology of CD-223 to CD4.

Specifications

Antigen LAG3
Applications Western Blot
Classification Monoclonal
Clone OTI8G6
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LAG3
Gene Accession No. P18627
Gene Alias CD223
Gene Symbols LAG3
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 66-332 of human LAG3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3902
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.