Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG3, Mouse anti-Human, Clone: 17B4, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16088
Specifications
LAG3 | |
Monoclonal | |
Unconjugated | |
In PBS (10% glycerol, 0.02% sodium azide). | |
CD223 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
17B4 | |
Immunohistochemistry Immunoprecipitation (1-2 ug/mL) Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
lymphocyte-activation gene 3 | |
LAG3 | |
A synthetic peptide corresponding to 30 amino acids at N-terminus of human LAG3. | |
RUO | |
3902 | |
For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction