Learn More
LAG3 Recombinant Rabbit Monoclonal Antibody (BLR027F), Bethyl Laboratories
Rabbit Recombit Monoclonal Antibody
$174.36 - $661.54
Specifications
| Antigen | LAG3 |
|---|---|
| Clone | BLR027F |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2648
|
Bethyl Laboratories, Inc
A700027T |
10 μL |
Each for $174.36
|
|
|||||
50-157-2647
|
Bethyl Laboratories, Inc
A700027 |
100 μL |
Each for $661.54
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommende
LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).Specifications
| LAG3 | |
| Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P18627 | |
| 3902 | |
| Between 475 and 525 | |
| Primary | |
| 4° C | |
| LAG3 |
| BLR027F | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
| Lymphocyte-activation gene 3, CD223 | |
| LAG3 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.