Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAGE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546520 Supplier Invitrogen™ Supplier No. PA546520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MRDADADAGG GADGGDGRGG HSCRGGVDTA AAPAGGAPPA HAPGPGRDAA Sequence homology: Human: 100%.

This gene belongs to the ESO/LAGE gene family, members of which are clustered together on chromosome Xq28, and have similar exon-intron structures. Unlike the other family members which are normally expressed only in testis and activated in a wide range of human tumors, this gene is ubiquitously expressed in somatic tissues. The latter, combined with the finding that it is highly conserved in mouse and rat, suggests that the encoded protein is functionally important. An intronless pseudogene with high sequence similarity to this gene is located on chromosome 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAGE3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LAGE3
Gene Accession No. Q14657
Gene Alias CVG5; DXS9879E; DXS9951E; EKC/KEOPS complex subunit LAGE3; ESO3; ITBA2; L antigen family member 3; L antigen family, member 3; LAGE3; Pcc1; Protein ESO-3; Protein ITBA2
Gene Symbols LAGE3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LAGE3 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8270
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.