Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAGE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557527
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557527 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557527 Supplier Invitrogen™ Supplier No. PA557527
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDADADAGGG ADGGDGRGGH SCRGGVDTAA APAGGAPPAH APGPGRDAAS Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 41%.

This gene belongs to the ESO/LAGE gene family, members of which are clustered together on chromosome Xq28, and have similar exon-intron structures. Unlike the other family members which are normally expressed only in testis and activated in a wide range of human tumors, this gene is ubiquitously expressed in somatic tissues. The latter, combined with the finding that it is highly conserved in mouse and rat, suggests that the encoded protein is functionally important. An intronless pseudogene with high sequence similarity to this gene is located on chromosome 9.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAGE3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAGE3
Gene Accession No. Q14657
Gene Alias CVG5; DXS9879E; DXS9951E; EKC/KEOPS complex subunit LAGE3; ESO3; ITBA2; L antigen family member 3; L antigen family, member 3; LAGE3; Pcc1; Protein ESO-3; Protein ITBA2
Gene Symbols LAGE3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LAGE3. Recombinant protein control fragment (Product #RP-95193).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8270
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.