Learn More
LAIR-1 (CD305) Mouse anti-Human, Brilliant Violet 605, Clone: DX26, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 744559
Description
The DX26 monoclonal antibody specifically binds to Leukocyte-associated immunoglobulin-like receptor 1 (LAIR-1/ hLAIR1/Leukocyte-associated Ig-like receptor 1) that is encoded by LAIR1, /and also known as, CD305. LAIR-1 is a ñ32 kDa type I transmembrane glycoprotein with a single immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory (ITIM) motifs. LAIR-1 is expressed on T cells, B cells, NK cells, monocytes, and dendritic cells. LAIR-1 recruits SHP-1 and SHP-2 tyrosine phosphatases upon activation. Crosslinking of the LAIR-1 expressed on T cells or NK cells can result in strong inhibition of cell-mediated cytotoxicity. Although it is structurally related to human killer cell inhibitory receptors, LAIR-1 does not appear to recognize MHC class I molecules and thus represents a novel MHC class I-independent mechanism of NK cell regulation.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
Specifications
LAIR-1 (CD305) | |
DX26 | |
Brilliant Violet 605 | |
CD305, LAIR1, hLAIR1, Leukocyte-associated Ig-like receptor 1 | |
Human NK Cell Clone | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.