Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAIR-1 (CD305) Mouse anti-Human, Brilliant Violet 711, Clone: DX26, BD Optibuild™

Catalog No. BDB744561 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744561 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744561 Supplier BD Biosciences Supplier No. 744561
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The DX26 monoclonal antibody specifically binds to Leukocyte-associated immunoglobulin-like receptor 1 (LAIR-1/ hLAIR1/Leukocyte-associated Ig-like receptor 1) that is encoded by LAIR1, /and also known as, CD305. LAIR-1 is a ñ32 kDa type I transmembrane glycoprotein with a single immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory (ITIM) motifs. LAIR-1 is expressed on T cells, B cells, NK cells, monocytes, and dendritic cells. LAIR-1 recruits SHP-1 and SHP-2 tyrosine phosphatases upon activation. Crosslinking of the LAIR-1 expressed on T cells or NK cells can result in strong inhibition of cell-mediated cytotoxicity. Although it is structurally related to human killer cell inhibitory receptors, LAIR-1 does not appear to recognize MHC class I molecules and thus represents a novel MHC class I-independent mechanism of NK cell regulation.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAIR-1 (CD305)
Classification Monoclonal
Clone DX26
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD305, LAIR1, hLAIR1, Leukocyte-associated Ig-like receptor 1
Host Species Mouse
Immunogen Human NK Cell Clone
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.