Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAIR1 Antibody (113), Alexa Fluor™ 594, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP143324AF594
Description
LAIR1 Monoclonal specifically detects LAIR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
LAIR1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
LAIR1 | |
The immunogen for this antibody was CD305. | |
Immunology | |
3903 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
113 | |
Western Blot, Flow Cytometry | |
CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
Armenian Hamster | |
0.1 mL | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction