Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAIR1 Antibody (342219), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2664PEATT594
Description
LAIR1 Monoclonal antibody specifically detects LAIR1 in Human samples. It is validated for Flow CytometrySpecifications
LAIR1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
342219 | |
Flow Cytometry | |
CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
Mouse myeloma cell line NS0-derived recombinant human LAIR1 isoform 1, Gln22-His163, Accession # Q6GTX8 | |
0.1 mL | |
Immunology | |
3903 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction