Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAIR1 Monoclonal Antibody (NKTA255)

Mouse Monoclonal Antibody

$626.50 - $995.00

Specifications

Antigen LAIR1
Clone NKTA255
Concentration 0.5 mg/mL
Content And Storage 4°C
Applications Flow Cytometry, Immunoprecipitation, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA133620
View Documents
Invitrogen™
MA133620
200 μL
Each for $626.50
Only null left
Add to Cart
 
PIMA133625
View Documents
Invitrogen™
MA133625
500 μg
Each for $995.00
Only null left
Edge
Add to Cart
 
Description

Description

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

LAIR1 (CD305) functions as an inhibitory receptor that plays a constitutive negative regulatory role on cytolytic function of natural killer (NK) cells, B-cells and T-cells. Activation by Tyr phosphorylation results in recruitment and activation of the phosphatases PTPN6 and PTPN11. It also reduces the increase of intracellular calcium evoked by B-cell receptor ligation. May also play its inhibitory role independently of SH2-containing phosphatases. Modulates cytokine production in CD4+ T-cells, down-regulating IL2 and IFNG production while inducing secretion of transforming growth factor beta. Down-regulates also IgG and IgE production in B-cells as well as IL8, IL10 and TNF secretion. Inhibits proliferation and induces apoptosis in myeloid leukemia cell lines as well as prevents nuclear translocation of NF-kappa-B p65 subunit/RELA and phosphorylation of I-kappa-B alpha/CHUK in these cells. Inhibits the differentiation of peripheral blood precursors towards dendritic cells.
Specifications

Specifications

LAIR1
0.5 mg/mL
Flow Cytometry, Immunoprecipitation, Western Blot
Unconjugated
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.2
Q6GTX8
3903
NK cells lines B12.100 and AM.25.
Antibody
NKTA255
4°C
Monoclonal
Liquid
IgG1
Human
LAIR1
5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; Lair-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1
LAIR1
Primary
Protein G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.