Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAIR1 Mouse anti-Human, Clone: 9.1C3, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2478000.2MG
Description
LAIR1 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
LAIR1 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready | |
CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
Mouse | |
40 kDa | |
0.2 mg | |
Primary | |
Recognizes a single protein of 40kDa, identified as the CD305 (LAIR1). It is an inhibitory receptor, expressed on the majority of peripheral mononuclear cells, including natural killer (NK) cells, T-cells, B-cells, monocytes, and dendritic cells. Highly expressed in naive T-cells and B-cells but no expression on germinal center B-cells. This MAb is expressed on CD34 positive hematopoietic progenitors. Inhibitory receptors regulate the immune response to prevent lysis of cells recognized as self. The gene is a member of both the immunoglobulin superfamily and the leukocyte-associated inhibitory receptor family. LAIR1 has been identified as an anchor for tyrosine phosphatase SHP-1, and may induce cell death in myeloid leukemias. Alternative splicing results in multiple transcript variants. This MAb is identical to LAIR-1, based on the fact that it not only precipitated a protein of 40kDa but it also binds specifically to LAIR-1 cDNA transfected COS7 cells as well as recognizes LAIR-1 fusion protein in ELISA. LAIR-1 expresses on hematopoietic progenitor, implicating its role in the regulation of hematopoiesis at early stage. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2 κ |
Flow Cytometry, Immunofluorescence, CyTOF | |
9.1C3 | |
Unconjugated | |
PBS with No Preservative | |
LAIR1 | |
Human Lymphocytes and Large Granular Lymphocytes (LGL) | |
Protein A or G purified | |
RUO | |
3903 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction