Learn More
Invitrogen™ LAIR1 Polyclonal Antibody

Description
Positive Samples: B-cell, Jurkat, HL-60, Raji, Mouse brain, Mouse liver, Mouse lung, Rat liver; Cellular Location: Cell membrane, Single-pass type I membrane protein Immunogen sequence: QEEDLPRPSI SAEPGTVIPL GSHVTFVCRG PVGVQTFRLE RDSRSTYNDT EDVSQASPSE SEARFRIDSV REGNAGLYRC IYYKPPKWSE QSDYLELLVK ESSGGPDSPD TEPGSSAGPT QRPSDNSHNE HAPASQGLKA EH.
Specifications
Specifications
| Antigen | LAIR1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.11 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LAIR1 |
| Gene Accession No. | Q6GTX8 |
| Gene Alias | 5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; Lair-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1 |
| Gene Symbols | LAIR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.