Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAIR2 Antibody (024), Alexa Fluor™ 532 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289685AF532
Description
LAIR2 Monoclonal antibody specifically detects LAIR2 in Human samples. It is validated for ELISASpecifications
LAIR2 | |
Monoclonal | |
Alexa Fluor 532 | |
CD306 antigen, CD306leukocyte-associated Ig-like receptor-2, LAIR-2, leukocyte-associated Ig-like receptor 2, leukocyte-associated immunoglobulin-like receptor 2, MGC71634 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human LAIR2 (NP_002279.2; Met 1-Pro 152). | |
0.1 mL | |
Immunology | |
3904 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
024 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction