Learn More
Lakeland Industries MicroMax™ Level C Coveralls
Catalog No. p-2638258
Encompass
Change view
Click to view available options
Size:
X-Large
Style:
Elastic wrists
Description:
Attached hood and boots, Composite Fabric
Catalog No. Size Style Description
19-146-311C X-Large Elastic wrists Attached hood and boots, Composite Fabric
1 options
This item is not returnable. View return policy
Catalog No. 19-146-311C Supplier Lakeland Industries Supplier No. TG414XL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Size X-Large
Material Spunbond Polypropylene, Nylon Scrim
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Description Attached hood and boots, Composite Fabric
Style Elastic wrists
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov