Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LALBA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576868
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576868 Supplier Invitrogen™ Supplier No. PA576868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes alpha-lactalbumin, a principal protein of milk. Alpha-lactalbumin forms the regulatory subunit of the lactose synthase (LS) heterodimer and beta 1,4-galactosyltransferase (beta4Gal-T1) forms the catalytic component. Together, these proteins enable LS to produce lactose by transfering galactose moieties to glucose. As a monomer, alpha-lactalbumin strongly binds calcium and zinc ions and may possess bactericidal or antitumor activity. A folding variant of alpha-lactalbumin, called HAMLET, likely induces apoptosis in tumor and immature cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LALBA
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LALBA
Gene Accession No. P00709, P00714
Gene Alias ALACTA; a-LACTA; alfaLA; alpha lactabumin; alpha lactalbumin; alpha-lactalbumin; alpha-lactalbumin A; alpha-lactalbumin B; alpha-lactalbumin precursor (EC 2.4.1.22); alpha-lactalbumin precursor protein; alpha-lactalbumin protein variant D; Bos d 4; lactalbumin; lactalbumin alpha; lactalbumin, alpha; lactalbumin, alpha-; lactalbumin, alpha- precursor; lactose synthase B protein; Lalba; lysozyme G; lysozyme-like protein 7; LYZG; LYZL7; MGC138521; MGC138523
Gene Symbols LALBA
Host Species Rabbit
Immunogen Recombinant full length Human LALBA.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24528, 3906
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.