Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMB1 Rabbit anti-Human, Unconjugated, Clone: AODF-12, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20439
Description
LAMB1 Monoclonal antibody specifically detects LAMB1 in Human samples. It is validated for WB-Ce, Flow Cyt.Specifications
LAMB1 | |
Monoclonal | |
Unconjugated | |
LAMB1 | |
Rabbit | |
100 μg | |
3912 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot, Flow Cytometry | |
AODF-12 | |
laminin, beta 1 | |
CLM, MGC142015 | |
A synthetic peptide corresponding to human LAMB1 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction