Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAMB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109899
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109899 Supplier Invitrogen™ Supplier No. PA5109899
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVRRAEQLLQ DARRARSWAE DEKQKAETVQ AALEEAQRAQ GIAQGAIRGA VADTRDTEQT LYQVQERMAG AERALSSAGE RARQLDALLE ALKLKRAGNS LAASTAEETA GSAQGRAQEA EQLLRGPLGD QYQTVKALAE RKAQGVLAAQ ARAEQLRDEA RDLLQAAQDK LQRLQELEGT YEENERALES KAAQLDGLEA RMRSVLQAIN LQVQIYNTCQ.

Nuclear lamins form a network of intermediate-type filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished, i.e. A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2. Recent evidence has revealed that mutations in A-type lamins give rise to a range of rare but dominant genetic disorders, including Emery-Dreifuss muscular dystrophy, dilated cardiomyopathy with conduction-system disease and Dunnigan-type familial partial lipodystrophy. In addition, the expression of A-type lamins coincides with cell differentiation and as A-type lamins specifically interact with chromatin, a role in the regulation of differential gene expression has been suggested for A-type lamins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMB2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LAMB2
Gene Accession No. P55268
Gene Alias AW211941; Lamb2; Lamb-2; Laminin B1s chain; laminin chain B3; laminin chain beta 2; Laminin S; laminin subunit beta 2; laminin subunit beta-2; laminin, beta 2; laminin, beta 2 (laminin S); laminin-11 subunit beta; Laminin-14 subunit beta; laminin-15 subunit beta; Laminin-3 subunit beta; laminin-4 subunit beta; Laminin-7 subunit beta; laminin-9 subunit beta; LAMS; NPHS5; npht; SLAM; S-LAM beta; S-laminin subunit beta
Gene Symbols LAMB2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1579-1798 of human LAMB2 (NP_002283.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3913
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.