Learn More
LAMB3 Mouse anti-Human, Clone: OTI1H10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806059

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The product encoded by this gene is a laminin that belongs to a family of basement membrane proteins. This protein is a beta subunit laminin, which together with an alpha and a gamma subunit, forms laminin-5. Mutations in this gene cause epidermolysis bullosa junctional Herlitz type, and generalized atrophic benign epidermolysis bullosa, diseases that are characterized by blistering of the skin. Multiple alternatively spliced transcript variants that encode the same protein have been found for this gene.Specifications
LAMB3 | |
Monoclonal | |
Unconjugated | |
LAMB3 | |
AI1A, BM600-125KDA, LAM5, LAMNB1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3914 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1H10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13751 | |
LAMB3 | |
Human recombit protein fragment corresponding to amino acids 22-315 of human LAMB3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.