Learn More
Invitrogen™ LAMB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109900
Description
Immunogen sequence: TGSPKLVALR LEMSSLPDLT PTFNKLCGNS RQMACTPISC PGELCPQDNG TACGSRCRGV LPRAGGAFLM AGQVAEQLRG FNAQLQRTRQ MIRAAEESAS QIQSSAQRLE TQVSASRSQM EEDVRRTRLL IQQVRDFLTD PDTDAATIQE VSEAVLALWL PTDSATVLQK MNEIQAIAAR LPNVDLVLSQ TKQDIARARR LQAEAEEARS RAHAVEGQVE DVVGNLRQGT VALQEAQDTM QGT.
The product encoded by this gene is a laminin that belongs to a family of basement membrane proteins. This protein is a beta subunit laminin, which together with an alpha and a gamma subunit, forms laminin-5. Mutations in this gene cause epidermolysis bullosa junctional Herlitz type, and generalized atrophic benign epidermolysis bullosa, diseases that are characterized by blistering of the skin. Multiple alternatively spliced transcript variants that encode the same protein have been found for this gene.
Specifications
| LAMB3 | |
| Polyclonal | |
| Unconjugated | |
| LAMB3 | |
| AI1A; BM600-125KDA; Epiligrin subunit bata; kalinin B1 chain; Kalinin subunit beta; kalinin-140kDa; LAM5; Lamb3; Laminin B1k chain; laminin beta 3; laminin chain; laminin S B3 chain; laminin subunit beta 3; laminin subunit beta-3; laminin, beta 3; laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)); laminin-5 subunit beta; LAMNB1; Nicein subunit beta; nicein, 125kD; nicein, 125kDa; nicein-125k; nicein-125kDa; RP1-272L16.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 305078, 3914 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q13751 | |
| LAMB3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 762-1004 of human LAMB3 (NP_0002192). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.