Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMB3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 267951AP150UL
Description
The product encoded by this gene is a laminin that belongs to a family of basement membrane proteins. This protein is a beta subunit laminin, which together with an alpha and a gamma subunit, forms laminin-5. Mutations in this gene cause epidermolysis bullosa junctional Herlitz type, and generalized atrophic benign epidermolysis bullosa, diseases that are characterized by blistering of the skin. Multiple alternatively spliced transcript variants that encode the same protein have been found for this gene.Specifications
| LAMB3 | |
| Polyclonal | |
| Unconjugated | |
| LAMB3 | |
| BM600 125KDA, Epiligrin subunit bata, Kalinin B1 chain, Kalinin subunit beta, LAM5, LAMB3, Laminin 5 subunit beta, Laminin B1k chain, Laminin subunit beta 3, laminin, beta 3, LAMNB1, Nicein subunit beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3914 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13751 | |
| LAMB3 | |
| LAMB3 Fusion Protein Ag25192 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction