Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS180P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
50μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS180P0 100 μL Concentrated 50μg/mL
MS180P1ABX 100 μL Concentrated 1mg/mL
MS180PABX 200 μL Concentrated 1mg/mL
MS180P 1 mL Concentrated 50μg/mL
MS180R1 1 mL Prediluted -
MS180R7 7 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS180P0 Supplier Epredia Supplier No. MS180P0

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Lambda Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: HP6054

Isotype: IgG2a/k

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Lambda Light Chain Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HP6054
Concentration 50μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human IgG myeloma proteins covalently coupled to polyaminostyrene micro beads
Quantity 100 μL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.