Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lambda Light Chain Antibody (1-155-2), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP179124JF646
Description
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.Specifications
LambdaLightChain | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGLC1 | |
Human IgA1 lambda myeloma protein | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody 1-155-2 recognizes lambda light chains (22.5 kDa) of human immunoglobulin. | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
1-155-2 | |
Flow Cytometry | |
IGLC, immunoglobulin lambda constant group | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Immunology | |
3536 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction