Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lambda Light Chain Antibody (4C2), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP145071PECY7
Description
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.Specifications
Lambda Light Chain | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
IGLC1 | |
The antibody 4C2 reacts with lambda light chains (22.5 kDa) of human immunoglobulin. | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody H4B4 recognizes CD107b/ LAMP-2, an extensively glycosylated 100-120 kDa widely expressed lysosome-associated protein. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
4C2 | |
Flow Cytometry | |
IGLC, immunoglobulin lambda constant group | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Immunology | |
3537.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction