Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lambda Light Chain Antibody (Mc24-IC6), Alexa Fluor™ 488

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063014AF488
Description
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
Lambda Light Chain | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IGLC1 | |
Purified human IgG. | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3536 | |
Human | |
Purified |
Flow Cytometry, ELISA | |
Mc24-IC6 | |
Flow Cytometry, ELISA | |
IGLC, immunoglobulin lambda constant group | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Recognises human lambda light chains, and shows no cross- reactivity to kappa light chains or to immunoglobulin heavy chains. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction