Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lambda Light Chain Antibody (Mc24-IC6), PerCP

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063014PCP
Description
Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.Specifications
| Lambda Light Chain | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| IGLC1 | |
| Purified human IgG. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3536 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| Mc24-IC6 | |
| Flow Cytometry | |
| IGLC, immunoglobulin lambda constant group | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognises human lambda light chains, and shows no cross- reactivity to kappa light chains or to immunoglobulin heavy chains. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction