Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lambda light chain Monoclonal Antibody (1-155-2), Super Bright™ 436, eBioscience™, Invitrogen™

Catalog No. 62999041
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-999-041 25 Tests
62-999-042 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 62-999-041 Supplier Invitrogen™ Supplier No. 62999041
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody 1-155-2 recognizes the lambda light chain of human immunoglobulin. Approximately 22-kDa, the Ig lambda chain is comprised of one variable and one constant region. Two lambda chains together with the heavy chain comprise the whole Ig structure. Expression of Ig lambda is typically found in less than half of B lymphocytes, and has been associated with neoplastic lymphomas. Applications Reported: This 1-155-2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1-155-2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lambda light chain
Applications Flow Cytometry
Classification Monoclonal
Clone 1-155-2
Concentration 5 μL/Test
Conjugate Super Bright 436
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IGLC1
Gene Accession No. P01701, P0CG04
Gene Alias Bence Jones Protein; BJP; Clambda1; Ig lambda chain V-I region BL2; IgL; Igl@; IGLC; IGLC 1/2/3; IGLC1; Igl-C1; IGLL5; IGLV; IGLV@; immmunoglobulin lambda light chain variable region; Immunoglobulin L; immunoglobulin lambda chain complex; immunoglobulin lambda constant 1; immunoglobulin lambda constant 7; immunoglobulin lambda light chain; immunoglobulin lambda light chain constant region; immunoglobulin lambda light chain V-J region; immunoglobulin lambda variable cluster; Lambda LC; lambda-immunoglobulin; LOC100060608; Mcg Marker; Paraprotein
Gene Symbols IGL, IGLC1, IGLV, IGLV1-51
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28820, 3535, 3537, 3546
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.