Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lambda light chain Monoclonal Antibody (5H1)
GREENER_CHOICE

Catalog No. PIMA549494
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549494 Supplier Invitrogen™ Supplier No. MA549494
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product was used in an Indirect ELISA.

Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lambda light chain
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene IGLV1-51
Gene Accession No. P01701, P0CG04
Gene Alias Bence Jones Protein; BJP; Clambda1; Ig lambda chain V-I region BL2; IgL; Igl@; IGLC; IGLC 1/2/3; IGLC1; Igl-C1; IGLL5; IGLV; IGLV@; immmunoglobulin lambda light chain variable region; Immunoglobulin L; immunoglobulin lambda chain complex; immunoglobulin lambda constant 1; immunoglobulin lambda constant 7; immunoglobulin lambda light chain; immunoglobulin lambda light chain constant region; immunoglobulin lambda light chain V-J region; immunoglobulin lambda variable cluster; Lambda LC; lambda-immunoglobulin; LOC100060608; Mcg Marker; Paraprotein
Gene Symbols IGL, IGLC1, IGLV, IGLV1-51
Host Species Mouse
Immunogen Recombinant Human Lambda.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28820, 3535, 3537, 3546
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.