Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB334R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
12 mL
Form:
Concentrated
Ready-to-Use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB334R1 1 mL Ready-to-Use
RB334A 1 mL Concentrated
RB334A0 100 μL Concentrated
RB334RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB334R1 Supplier Epredia Supplier No. RB334R1

One marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available is the Epredia™ Lambda Light Chain, Rabbit Polyclonal Antibody.

Antibody to the λ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Baboon, Mouse and Rat. Does not react with dog. Others not known.

Epitope: Not determined

Immunogen: Lambda light chains purified from human Bence Jones proteins

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Lambda Light Chain
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Lambda light chains purified from human Bence Jones proteins
Quantity 1 mL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Baboon, Human, Mouse, Rat
Form Ready-to-Use
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.