Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lambda5/IGLL1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299452MFV450
Description
Lambda5/IGLL1 Polyclonal antibody specifically detects Lambda5/IGLL1 in Mouse samples. It is validated for Western BlotSpecifications
Lambda5/IGLL1 | |
Polyclonal | |
Western Blot | |
14.1, AGM2, CD179 antigen-like family member B, CD179b, CD179b antigen, Ig lambda-5, IGL1, IGL5CD179B, IGLJ14.1, IGLL, IGO, IGVPB, immunoglobulin lambda-like polypeptide 1, Immunoglobulin omega polypeptide, immunoglobulin omega polypeptide chain, immunoglobulin-related 14.1 protein, Immunoglobulin-related protein 14.1, lambda5, Pre-B lymphocyte-specific protein-2, VPREB2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Lambda5/IGLL1 | |
0.1 mL | |
Immunology | |
3543 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction