Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30516720UL
Description
LAMC2 Polyclonal specifically detects LAMC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
LAMC2 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
3918 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
BM600, BM600-100kDa, Cell-scattering factor 140 kDa subunit, CSF, CSF 140 kDa subunit, EBR2, EBR2A, Epiligrin subunit gamma, kalinin (105kD), Kalinin subunit gamma, Kalinin/nicein/epiligrin 100 kDa subunit, kalinin-105kDa, ladsin (140kDa), Ladsin 140 kDa subunit, LAMB2Tlaminin, gamma 2 (nicein (100kD), kalinin (105kD), BM600 (100kD), Herlitzjunctional epidermolysis bullosa)), Laminin B2t chain, laminin subunit gamma-2, laminin, gamma 2, Laminin-5 subunit gamma, LAMNB2B2T, Large adhesive scatter factor 140 kDa subunit, MGC138491, MGC141938, nicein (100kDa), Nicein subunit gamma, nicein-100kDa | |
Recombinant fusion protein containing a sequence corresponding to amino acids 862-1111 of human LAMC2 (NP_061486.2). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction