Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LAMC3 |
---|---|
Concentration | 0.45 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2663
![]() ![]() |
Proteintech Group Inc
200301AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to LAMC3.
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kDa) and three short arms (200 kDa). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.Specifications
LAMC3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9R0B6, Q9Y6N6 | |
10319, 23928 | |
Peptide | |
Primary | |
-20°C | |
LAMC3 |
0.45 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
LAMC3, Laminin 12 subunit gamma, Laminin 14 subunit gamma, Laminin 15 subunit gamma, Laminin subunit gamma 3, laminin, gamma 3 | |
LAMC3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title