Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin A + C Antibody (133A2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197688F
Description
Lamin A + C Monoclonal specifically detects Lamin A + C in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Lamin A | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
LMNA | |
133A2 is a mouse monoclonal IgG3/kappa antibody obtained from fusion of P3/X63.Ag8.653 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with partially purified recombinant human lamin A. | |
0.1 mL | |
Primary | |
133A2 recognizes an epitope located between residues 598-611 of lamin A and therefore 133A2 reacts exclusively with lamin A. | |
Store at 4C in the dark. | |
IgG3 |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
133A2 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32 | |
Mouse | |
Protein A or G purified | |
RUO | |
4000 | |
Human, Mouse, Rat, Bovine, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction