Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin A + C Antibody (4C4), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$206.50 - $501.50
Specifications
| Antigen | Lamin A + C |
|---|---|
| Clone | 4C4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:1000 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP225151SS
![]() |
Novus Biologicals
NBP225151SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP225151
![]() |
Novus Biologicals
NBP225151 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
Lamin A + C Monoclonal specifically detects Lamin A + C in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Lamin A + C | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32 | |
| LMNA | |
| IgG1 | |
| Affinity Purified | |
| Human, Bovine, Porcine, Mouse and Rat. Other species have not been tested. |
| 4C4 | |
| Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:1000 | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat, Monkey | |
| P02545 | |
| 4000 | |
| Full length recombinant human lamin A expressed in and purified from E. coli. [UniProt# P02545] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 74/65 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title