Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A + C Antibody, Novus Biologicals™
SDP

Catalog No. NBP225152SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225152SS 0.025 mL
NBP225152 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225152SS Supplier Novus Biologicals Supplier No. NBP225152SS
Only null left
Add to Cart
Add to Cart

Chicken Polyclonal Antibody has been used in 1 publication

Lamin A + C Polyclonal specifically detects Lamin A + C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Lamin A + C
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:1000
Formulation Supplied as concentrated IgY in PBS with 5mM Sodium Azide
Gene Accession No. P02545
Gene Alias CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32
Gene Symbols LMNA
Host Species Chicken
Immunogen Full length recombinant human lamin A protein expressed in and purified from E. coli.
Molecular Weight of Antigen 74/65 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4000
Test Specificity Human, Bovine, Porcine, Mouse, Rat and other Mammals. Other species have not been tested.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.