Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A/C Monoclonal Antibody (3A6-4C11), Active Motif™
SDP

Catalog No. 501993990
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
50 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3990 10 μg
50-199-3989 100 μg
50-227-7232 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-3990 Supplier Active Motif Supplier No. 39288
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).

Specifications

Antigen Lamin A/C
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3A6-4C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation antibody with 30% glycerol and 0.035% sodium azide; pH
Gene LMNA
Gene Accession No. P02545, P48678, P48679
Gene Alias 70 kDa lamin; BOS_2775; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; I79_009616; IDC; lamin; lamin A; lamin A/C; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; lamin C; lamin C2; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; LMNA; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; Prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1
Gene Symbols LMNA
Host Species Mouse
Immunogen This Lamin A/C antibody was raised against a recombinant protein corresponding to amino acids 430-545 of human Lamin A.
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100757316, 16905, 4000, 60374
Target Species Hamster, Human, Monkey, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.