Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lamin A/C, Mouse, Unlabeled, Clone: 2E8.2,

Catalog No. MABT538MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT538MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT538MI Supplier MilliporeSigma™ Supplier No. MABT538
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lamin-A is a key component of the nuclear lamina, a protein network underlying the inner membrane that determines nuclear shape and size. The same gene (Lmna, Dhe, or Lamn1 in murine species; Gene ID 16905) that encodes lamin-A also codes for the alternatively spliced prelamin-C isoforms (UniProt P48678-2 and P48678-3). Murine lamin-A (UniProt P48678-1) is initially produced as a 665-amino acid prelamin A, which is further processed posttranslationally by four enzymatic steps to yield the mature Lamin-A. Farnesylation of Cys662, endoproteolytic cleavage of the last three amino acids (a.a. 663-665), carboxyl methylation of Cys662, and the final endoproteolytic removal of the remaining propeptide sequence (a.a. 648-662), including the farnesylated Cys662, catalyzed by the ER membrane zinc metalloproteinase ZMPSTE24.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamin A/C
Applications Western Blot
Classification Monoclonal
Clone 2E8.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene LMNA, LMN1
Gene Accession No. P02545
Host Species Mouse
Immunogen GST-tagged recombinant fragment within the C-terminal half of human Lamin A.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_733821
Test Specificity Clone 2E8.2 immunostained nuclear membrane in human tissue sections.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.