Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Lamin A/C Polyclonal Antibody
Chicken Polyclonal Antibody
$533.50 - $1763.00
Specifications
Antigen | Lamin A/C |
---|---|
Concentration | Conc. Not Determined |
Content And Storage | 4°C |
Applications | Western Blot, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5143540
|
Invitrogen™
PA5143540 |
500 μL |
Each for $1,763.00
|
|
|||||
PIPA5143574
|
Invitrogen™
PA5143574 |
100 μL |
Each for $533.50
|
|
|||||
Description
Lamin A/C Polyclonal Antibody for Western Blot, ICC/IF
Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).Specifications
Lamin A/C | |
4°C | |
Polyclonal | |
Liquid | |
IgY | |
Human, Mouse, Rat | |
LMNA | |
70 kDa lamin; cb948; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; I79_009616; IDC; lamin; lamin A; lamin A/C; lamin A/C L homeolog; lamin A/C-like 1; Lamin A+C mutant; Lamin AC; lamin a-c; lamin C; lamin C2; lamin-A; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; lmna; LMNA protein; lmna.L; lmna-A; LMNC; LMNL1; mutant 453W; Mutant lamin A/C; nuclear intermediate filament protein; nuclear lamin A; prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32; RP11-54H19.1; unnamed protein product; wu:fk66d12; XELAEV_18040808mg | |
LMNA | |
Primary |
Conc. Not Determined | |
Western Blot, Immunocytochemistry, Western Blot | |
Unconjugated | |
Chicken | |
RUO | |
PBS with 0.02% sodium azide | |
P02545, P48678, P48679 | |
16905, 4000, 60374 | |
Full length recombinant human lamin A protein expressed in and purified from E. coli. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title