Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin A + C R453W Antibody (12A-2F5), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP177401AF700
Description
Lamin A + C R453W Monoclonal specifically detects Lamin A + C R453W in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Lamin A + C R453W | |
Monoclonal | |
Alexa Fluor 700 | |
NP_733821.1 | |
LMNA | |
Synthetic peptide corresponding to amino acids 449-457 of human Lamin A + C [Accession#: NP_733821.1] | |
0.1 mL | |
Cancer, Chromatin Modifiers, Chromatin Research, DNA replication Transcription Translation and Splicing, Nuclear Envelope Markers, Signal Transduction | |
4000 | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
12A-2F5 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
70 kDa Lamin, CDCD1, CDDC, Charcot Marie Tooth Disease Axonal Type 2B1, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LAMIN A, Lamin A/C, Lamin A/C R453W, LAMIN C, LDP1, LFP, LGMD1B, LMN 1, LMN A, LMN C, LMN1, LMNA, LMNC, PRELAMIN A, PRO1, Progeria 1 (Hutchinson Gilford Type) | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Disease-associated point-mutant R453W of Lamin A + C | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction