Learn More
Invitrogen™ Lamin A (Cleaved Asp230) Monoclonal Antibody (C.69.9)

Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with full length lamin A or C.
Specifications
Specifications
| Antigen | Lamin A (Cleaved Asp230) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | C.69.9 |
| Concentration | 1 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 |
| Gene | LMNA |
| Gene Accession No. | P02545, P48678, P48679 |
| Gene Alias | 70 kDa lamin; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; lamin; lamin A; lamin A/C; lamin A/C-like 1; lamin C; lamin C2; lamin-A/C; LDP1; LFP; LGMD1B; Lmn1; LMNA; LMNC; LMNL1; OTTHUMP00000015843; OTTHUMP00000015846; OTTHUMP00000015847; Prelamin-A/C; PRO1; Renal carcinoma antigen NY-REN-32 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.