Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin B Mouse anti-Human, Clone: 9C11, Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM33040M

Description
Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Laminins are composed of 3 non identical chains: laminin alpha, beta and gamma (formerly A, B1, and B2, respectively) and they form a cruciform structure consisting of 3 short arms, each formed by a different chain, and a long arm composed of all 3 chains. Defects in the LMNB1 gene can cause Leukodystrophy, demyelinating, autosomal dominant, adult-onset (ADLD).Specifications
Lamin B | |
Monoclonal | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P20700 | |
Lmnb1 | |
Recombinant human Lamin B Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
9C11 | |
Unconjugated | |
Lmnb1 | |
ADLD; BOS_7828; fc06g01; I79_003938; lamin B1; lamin B1 L homeolog; lamin B1 protein; lamin-b; Lamin-B1; Lamin-L(I); LMN; LMN2; LMNB; Lmnb1; lmnb1 protein; lmnb1.L; MGC111419; wu:fc06g01; XELAEV_18007951mg | |
Mouse | |
Protein G | |
RUO | |
4001 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction