Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B Receptor (LBR), Mouse anti-Human, Mouse, Clone: BB2SS3F3, Millipore Sigma™

Catalog No. MABT83125UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT83125UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT83125UG Supplier MilliporeSigma Supplier No. MABT83125UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Lamin B Receptor (LBR), clone BB2SS3F3, Cat. No. MABT831, is a mouse monoclonal antibody that detects Lamin B receptor and has been tested for use in Immunocytochemistry, Immunofluorescence, and Western Blotting.

Lamin-B receptor (UniProt: Q14739; also known as Integral nuclear envelope inner membrane protein, LMN2R) is encoded by the LBR gene (Gene ID: 3930) in human. Lamin-B receptor is a multi-pass membrane protein that anchors the lamina and the heterochromatin to the inner nuclear membrane. It is an integral membrane protein of the interphase nuclear envelope (NE). It contains a Tudor damain that lies within the first 62 amino acids. Its N-terminal end resides in the nucleoplasm, binding to lamin B and heterochromatin and the C-terminal end resides within the inner nuclear membrane, retreating with the endoplasmic reticulum (ER) away from condensing chromosomes during mitotic NE breakdown. During mitosis, Lamin-B receptor is retained within the retreating ER and kept out of the way of the mitotic apparatus. Lamin- B receptor is suggested to perform three vital functions in the cell: (a) contributes to post-mitotic nuclear reformation; (b) contributes to interphase NE growth; (c) contributes to compartmentalization of heterochromatin within the interphase nucleus. Lamin-B receptor is phosphorylated by Cdk1 in mitosis when the inner nuclear membrane breaks down into vesicles that dissociate from the lamina and the chromatin. It is phosphorylated by different protein kinases in interphase when the membrane is associated with these structures. Mutations in LBR gene are known to cause Pelger-Huet anomaly that is characterized by skeletal anomalies, developmental delay, and seizures. Some mutations cause Greenberg dysplasia that can lead to in utero lethality. (Ref.: Olins, AL., et al. (2010). Nucleus 1(1); 53-70).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamin B Receptor (LBR)
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone BB2SS3F3
Conjugate Unconjugated
Dilution Immunofluorescence Analysis: A representative lot detected Lamin B Receptor (LBR) in C57BL/6 mouse small intestine (Courtesy of Dr. Brian Burke, Institute of Medical Biology, A*STAR, Singapore).Western Blotting Analysis: A representative lot detected Lamin B Receptor (LBR) in HeLa cell lysate, mouse C2C12 cell lysate, and in adult mouse fibroblasts (MAFs) either LBR+/+ or LBR -/- (Courtesy of Dr. Brian Burke, Institute of Medical Biology, A*STAR, Singapore).Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected Lamin B Receptor (LBR) in HeLa cells.Immunocytochemistry Analysis: A representative lot detected Lamin B Receptor (LBR) in HeLa and NIE-115 cells, as well as in mouse adult fibroblasts (MAF LBR+/+ vs MAF LBR-/-) (Courtesy of Dr. Brian Burke, Institute of Medical Biology, A*STAR, Singapore).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias LMN2R;Integral nuclear envelope inner membrane protein;LBR
Gene Symbols LBR
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to the first 211 amino acids from human Lamin B receptor.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone BB2SS3F3 detects Lamin B receptor in human cells. It targets an epitope within the first 211 amino acids.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.