Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B Receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986318
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6318 Supplier Bioss Supplier No. BS5081R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the ERG4/ERG24 family. It localized in the nuclear envelope inner membrane and anchors the lamina and the heterochromatin to the membrane. It may mediate interaction between chromatin and lamin B. Mutations of this gene has been associated with autosomal recessive HEM/Greenberg skeletal dysplasia. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.

Specifications

Antigen Lamin B Receptor
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LBR
Gene Accession No. Q14739
Gene Alias 3-beta-hydroxysterol Delta (14)-reductase; AI505894; BOS_15656; C-14 sterol reductase; C14SR; Delta(14)-sterol reductase LBR; Delta-14-SR; DHCR14B; FLJ43126; ic; ichthyosis; Integral nuclear envelope inner membrane protein; lamin B receptor; lamin B receptor-like; Lamin-B receptor; LBR; LMN2R; LOC100345014; MGC9041; NBP60; PHA; PRO0650; sb:cb406; Sterol C14-reductase; TDRD18; tudor domain containing 18; wu:fb75h08; wu:fc47b04; wu:fd36b07; zgc:86649
Gene Symbols LBR
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Lamin B Receptor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3930
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.