Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lamin B1 Monoclonal Antibody (C4), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA550963
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550963 200 μL
FEMA550964 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550963 Supplier Invitrogen™ Supplier No. MA550963
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lamin B1 is a key member of the nuclear lamina structure, encoded by the LMNB1 gene. It is part of the B-type lamins, ubiquitously expressed across all mammalian cells, providing structural integrity and mechanical support by forming a meshwork along the inner nuclear membrane. Lamin B1 is involved in maintaining the shape and stability of the nucleus, playing a critical role in cellular processes such as genome organization, DNA replication, and transcription regulation. The lamin filaments are type V intermediate filament proteins, enabling the nucleus to withstand mechanical stress and preserve its functionality. Lamin B1's expression and dynamics vary during cell differentiation, influencing gene regulation. Its location within the euchromatin-enriched section of the genome suggests potential roles in chromatin organization and the epithelial-to-mesenchymal transition. Governed by complex interactions with other proteins, lamin B1 is integral to cellular architecture and health, with variations in its levels linked to developmental and disease processes. Recent studies have highlighted the importance of understanding the role and dynamics of lamin B1 in human-induced pluripotent stem cells and germ layer cells, offering insights into differentiation-dependent changes and their implications for nuclear structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamin B1
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Lmnb1
Gene Accession No. P20700
Gene Alias 0710005A05Rik; ADLD; C-flanking peptide of NPY; CPON; fc06g01; I79_003938; lamin B1; lamin B1 L homeolog; lamin B1 protein; lamin-b; lamin-B1; Lamin-L(I); LMN; LMN2; LMNB; LMNB1; lmnb1 protein; lmnb1.L; MGC111419; Neuropeptide tyrosine; Neuropeptide Y; NPY; NPY02; prepro-neuropeptide Y; pro-neuropeptide Y; PYY4; RATNPY; RATNPY02; unnamed protein product; wu:fc06g01; XELAEV_18007951mg
Gene Symbols Lmnb1
Host Species Mouse
Immunogen Recombinant Human LMNB1 protein, Ser52-Gly233 (Accession #P20700).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100346094, 4001
Target Species Human, Rabbit
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.