Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B1 Mouse anti-Human, CoraLite 555, Clone: 3C10G12, Proteintech
SDP

Catalog No. 502257444 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-7444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-7444 Supplier Proteintech Group Inc Supplier No. CL55566095100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Lamin B1 antibody from Proteintech is a mouse monoclonal antibody to a fusion protein of human Lamin B1. This antibody recognizes human, mouse, rat antigen. The Lamin B1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™555.

Specifications

Antigen Lamin B1
Applications Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 3C10G12
Conjugate CoraLite 555
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 4001
Target Species Human, Mouse, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.