Learn More
Invitrogen™ Lamin B1 Recombinant Rabbit Monoclonal Antibody (10H34L18)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ 702972

Description
This antibody is predicted to react with Monkey, Sheep, Bovine, Pig. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Lamin B1 is a key member of the nuclear lamina structure, encoded by the LMNB1 gene. It is part of the B-type lamins, ubiquitously expressed across all mammalian cells, providing structural integrity and mechanical support by forming a meshwork along the inner nuclear membrane. Lamin B1 is involved in maintaining the shape and stability of the nucleus, playing a critical role in cellular processes such as genome organization, DNA replication, and transcription regulation. The lamin filaments are type V intermediate filament proteins, enabling the nucleus to withstand mechanical stress and preserve its functionality. Lamin B1's expression and dynamics vary during cell differentiation, influencing gene regulation. Its location within the euchromatin-enriched section of the genome suggests potential roles in chromatin organization and the epithelial-to-mesenchymal transition. Governed by complex interactions with other proteins, lamin B1 is integral to cellular architecture and health, with variations in its levels linked to developmental and disease processes. Recent studies have highlighted the importance of understanding the role and dynamics of lamin B1 in human-induced pluripotent stem cells and germ layer cells, offering insights into differentiation-dependent changes and their implications for nuclear structure.
Specifications
Lamin B1 | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P14733, P20700, P70615 | |
Lmnb1 | |
Peptides corresponding to human LAMIN B1 [(aa62-aa79) (aa450-aa467)]. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
10H34L18 | |
Unconjugated | |
Lmnb1 | |
0710005A05Rik; ADLD; C-flanking peptide of NPY; CPON; fc06g01; I79_003938; lamin B1; lamin B1 L homeolog; lamin B1 protein; lamin-b; lamin-B1; Lamin-L(I); LMN; LMN2; LMNB; LMNB1; lmnb1 protein; lmnb1.L; MGC111419; Neuropeptide tyrosine; Neuropeptide Y; NPY; NPY02; prepro-neuropeptide Y; pro-neuropeptide Y; PYY4; RATNPY; RATNPY02; unnamed protein product; wu:fc06g01; XELAEV_18007951mg | |
Rabbit | |
Protein A | |
RUO | |
116685, 16906, 4001 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.