Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin B2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24888225UL
Description
Lamin B2 Polyclonal antibody specifically detects Lamin B2 in Human samples. It is validated for Western BlotSpecifications
Lamin B2 | |
Polyclonal | |
Western Blot 0.04-0.4 μg/mL | |
lamin B2, lamin-B2, LMN2LAMB2, MGC2721 | |
This antibody was developed against a recombinant protein corresponding to amino acids: DEVNKSAKKREGELTVAQGRVKDLESLFHRSEVELAAALSDKRGLESDVAELRAQLAKAEDGHAVAKKQLEKETLM | |
25 μL | |
Apoptosis, Cell Biology, Cytoskeleton Markers | |
84823 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction