Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin B2 Antibody (OTI2C6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272303F
Description
Lamin B2 Monoclonal antibody specifically detects Lamin B2 in Human samples. It is validated for Western BlotSpecifications
Lamin B2 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
OTI2C6 | |
Western Blot | |
lamin B2, lamin-B2, LMN2LAMB2, MGC2721 | |
Full length human recombinant protein of human LMNB2 (NP_116126) produced in HEK293T cell. | |
0.1 mL | |
Apoptosis, Cell Biology, Cytoskeleton Markers | |
84823 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction